Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Acid-Treated Low-Metal TCLP Filters

Catalog No. p-108752
Click to view available options
Diameter (Metric):
47 mm
90 mm
110 mm
125 mm
142 mm
150 mm

Binder free filters

The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6μm to 0.8μm

  • Resistant to acids and bases
  • Available in six diameters, including sizes to fit Büchner funnels and zero-headspace extraction units
  • Pretreatment with acid eliminates need to acid-wash filters in your lab, for reduced analysis time and reagent costs

Compliance

Conform to requirements of U.S. EPA Method 1311 Toxicity Characteristic Leaching Procedure (TCLP), including 0.6 to 0.8μm particle retention requirement.

TRUSTED_SUSTAINABILITY

Specifications

Description GE’s Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action.
Format Disc
Material Borosilicate Glass
Particle Retention 0.7 μm
For Use With (Application) Required for volatile samples and use with a zero headspace extractor.
Quantity 50 pieces
Shape Circle
Thickness (Metric) 420 μm
Type TCLP Limits Test Filter
Waterflow Rate 60 mL/min
Diameter (English) 3.5 in.
Diameter (Metric) 90 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.