Learn More
GE Healthcare Whatman™ Adsorption 60Å Silica Gel TLC Plates
Provide quality separation of moderately to strongly polar compounds, including carbohydrates, antibiotics, proteins, amino acids, and phospholipids
Manufacturer:  GE Healthcare 4860820
 View more versions of this product
Catalog No. 05-713-318
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 250µm layer thickness
  • 2.5 x 7.6cm size is a microscope slide plate
  • With organic binder and moderate layer hardness for convenient spot recovery with a sample recovery tube
  • Versions with Linear-K preadsorbent strip have a 3cm strip of inert material that acts as an inactive blotter or staging area for rapid application of samples along the edge of standard silica plate
Specifications
Specifications
20 x 20 cm
Partisil K6 60A