Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Anotop™ 25 Sterile Syringe Filters
Remove virtually 100% of particulates larger than pore size
Supplier: Cytiva 68092122
Description
Remove virtually 100% of particulates larger than pore size
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes 100mL (25mm diameter filter).
- Nondeformable, honeycombed Anopore™ membrane
- Inherently hydrophilic membrane is free of adhesives and surfactants
- Low protein binding, low fluid retention for maximum sample recovery
- Unidirectional flow
Order Info
Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders. Sterile units are individually packaged to avoid contamination.
Specifications
Anotop 25 mm Sterile Syringe Filters, 0.2 μm (50 pcs) | |
Anapore | |
Sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene (PP) | |
Female luer lock | |
6.9 bar (100 psi) | |
113°F | |
Polypropylene | |
100 mL (25 mm Diameter Filter) |
Single-use | |
0.2 μm | |
4.78 cm2 | |
<150 μL | |
Hydrophilic | |
Male luer | |
50/Pk. | |
45°C | |
Syringe Filter | |
25 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction