Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Anotop™ 25 Sterile Syringe Filters Available on GSA/VA Contract for Federal Government customers only.

Remove virtually 100% of particulates larger than pore size

Manufacturer:  GE Healthcare 68092112

 View more versions of this product

Catalog No. 09-926-12


Add to cart

Description

Description

Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes 100mL (25mm diameter filter).

  • Nondeformable, honeycombed Anopore™ membrane
  • Inherently hydrophilic membrane is free of adhesives and surfactants
  • Low protein binding, low fluid retention for maximum sample recovery
  • Unidirectional flow

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders. Sterile units are individually packaged to avoid contamination.

Specifications

Specifications

Anotop 25 mm Sterile Syringe Filters, 0.1 μm (50 pcs)
Syringe Filter
Anapore
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Polypropylene (PP)
Yes
Female luer lock
113°F
50/Pk.
Polypropylene
Sterile
25 mm
6.9 bar (100 psi)
<150μL
Whatman Anotop sterile syringe filters from GE contain an inorganic Anopore membrane. The low protein binding exhibited by this sterile membrane filter make it well suited for biological filtration.
4.78 cm2
Male luer
45°C
0.1μm
5.2kg/cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit