Learn More
GE Healthcare Whatman™ Anotop 10mm HPLC Syringe Filters, Pore size 0.2µm Available on GSA/VA Contract for Federal Government customers only.
Whatman Anotop HPLC syringe filter products from GE Healthcare’s Life Sciences business are specifically designed for effective UHPLC and HPLC sample preparation.
Manufacturer:  GE Healthcare 20010100
 View more versions of this product
Catalog No. 09-927-387
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Syringe filter removes particulates that could cause damage, supporting longer column life
  • Low levels of UV-absorbing extractable substances for consistent HPLC sample preparation
  • Anopore inorganic aluminum membrane filters a broad range of solvents
  • High pore-size precision due to capillary pore structure
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Non-sterile
1cm
100mL
0.2μm
Syringe Filter
Proprietary Alumina Based Anapore Membrane
100/Pk.