Learn More
GE Healthcare Whatman™ Application Specific Filters Available on GSA/VA Contract for Federal Government customers only.
Acid-washed filter for the determination of trace elements (Mg, Mn, Co, Cu, Mo, B).
Manufacturer:  GE Healthcare 10310643
 View more versions of this product
Catalog No. 09-924-146
Add to cart
Description
Description
Whatman offers a line of cellulose filter papers for specific applications. The product line includes filter papers for use in soil analysis and filter papers for the sugar industry.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
For use in soil analysis, filter papers for sugar industry
110mm
Filter; Grade 512.5