Learn More
GE Healthcare Whatman™ Solvent IFD, 0.2µm, 50mm, polypropylene Available on GSA/VA Contract for Federal Government customers only.
GE’s Solvent IFD is a high-flow HPLC in-line filter/degasser, designed to simultaneously filter and degas organically based HPLC mobile phases.
Manufacturer:  GE Healthcare 67255002A
 View more versions of this product
Catalog No. 09-800-984
Add to cart
Description
Description
  • Faster than traditional mobile phase preparation
  • Chemically resistant polypropylene membrane and housing, built to filter solvent mobile phases without expensive stand-alone degassing equipment
  • Air vent on inlet with luer lock cap
  • Integrity-testable with the bubble point method
  • End fittings included – for replacement rings and caps, see our IFD end fitting kit
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
50mm
Polypropylene (PP)
Connections: 1/8 in. tubing only
16cm2
0.2μm
10 pieces
Whatman Aqueous and Solvent IFD Inline Filter Degassers
Solvent IFD
Documents
Promotions
Promotions
  • GE Healthcare Life Sciences , Fisherbrand
    Get 5% off your purchase of $500 to $999.99 in GE Healthcare™ Life Sciences products with promo code GESAVE5.
    : GESAVE5
    09-30-2018