Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Autovial™ Syringeless Filters: 12mL Capacity Available on GSA/VA Contract for Federal Government customers only.

Faster sample handling, up to 12mL in volume

Manufacturer:  GE Healthcare AV125SAQU

 View more versions of this product

Catalog No. 09-916C


Add to cart

Description

Description

Replace syringe-coupled devices with single filter units for one-time use. Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.

  • Preassembled units consist of a filter barrel and a plunger
  • Built-in air purge minimizes holdup
  • Integrated construction eliminates filter pop-off
  • Support stand protects slip-Luer tip, recessed to reduce chance of contamination
  • Handle samples up to 12mL in volume
  • All have an integral 25mm diameter filter in barrel (graduated in 1mL intervals) with polyethylene plunger
  • Filtration area: 3.0cm2
  • Glass microfiber prefilter ensures higher flow with less clogging
  • Units with NYL, PTFE, and PVDF membranes are available sterile

Cellulose Acetate (CA)

  • Low nonspecific protein binding, high loading capacity; for biologicals
Glass Microfiber (GMF)
  • High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
Nylon (NYL)
  • Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
Polypropylene (PP)
  • For aqueous and organic solutions; ideal for aggressive samples and high debris
Polytetrafluoroethylene (PTFE)
  • Hydrophobic; for samples >50% organic solvents
Polyvinylidene Fluoride (PVDF)
  • Low protein binding; for aqueous samples

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Autovial 12 Syringeless Filter, glass prefilter, sterile, 0.2 μm, PVDF filtration medium (40 pcs)
Syringeless Filter
PTFE
140 μl
The Whatman Autovial Syringeless Filter replaces multiple syringe-coupled sample filtration devices with a single, disposable unit.
Yes
Male slip luer
0.2μm
Sterile
25mm
Sample preparation, Difficult-to-filter samples, Quick filtration of samples, Low nonspecific protein binding membrane for samples in aqueous solutions and/or organic solvents.
Polypropylene (PP)
<=12mL
3 cm2
40/Pk.
Polypropylene
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit