Learn More
Cytiva Whatman™ Autovial™ 5mL Syringeless Filters
Replaces syringe-coupled devices with single filter units for one-time use. Whatman™ Autovial™ 5mL Syringeless Filters are pre-assembled filtration devices for removing particulates from samples.
$386.00 - $405.00
Specifications
Sterility | Non-sterile |
---|---|
Quantity | 50/Pk. |
Type | Syringeless Filter |
Volume (Metric) | ≤5 mL |
Diameter (Metric) | 13 mm |
Catalog Number | Mfr. No. | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Pore Size | Quantity | Price | Quantity & Availability | |||
09-921-13
|
Cytiva
AV115NPEORG |
PTFE | 0.2 μm | 50/Pk. |
Pack of 50 for $386.00
|
|
|||
09-921-15
|
Cytiva
AV115NPUAQU |
PVDF | 0.45 μm | 50/Pk. |
Pack of 50 for $395.00
|
|
|||
09-921-17
|
Cytiva
AV115NPUNYL |
Nylon | 0.45 μm | 50/Pk. |
Pack of 50 for $405.00
|
|
|||
09-921-18
|
Cytiva
AV115NPUORG |
PTFE | 0.45 μm | 50/Pk. |
Pack of 50 for $394.00
|
|
|||
09-921-20
|
Cytiva
AV115UGMF |
Glass Microfiber | 0.45 μm | 50/Pk. |
Pack of 50 for $394.00
|
|
|||
Description
Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Support stand protects slip-Luer tip, recessed to reduce chance of contamination
- Remove particulates from samples up to 5mL in volume
- Filtration area: 1.7cm2
Glass Microfiber (GMF)
- High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
- Nylon (NYL)
- Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
- Polytetrafluoroethylene (PTFE)
- Hydrophobic; for samples >50% organic solvents
- Polyvinylidene Fluoride (PVDF)
- Low protein binding; for aqueous samples
Specifications
Non-sterile | |
Syringeless Filter | |
13 mm |
50/Pk. | |
≤5 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.