Learn More
Cytiva Whatman™ Autovial Syringeless Filters::Autovial 12::Without Prefilter
For removing particulates from potentially hazardous samples
$555.00
Specifications
Pore Size | 0.45 μm |
---|---|
Filtration Area | 3 cm2 |
Hold up Volume (After Air Purge) | 140 μL |
Housing Material | Polypropylene (PP) |
Outlet Style | Male slip luer |
Description
Support stand protects slip-Luer tip, recessed to reduce chance of contamination. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial or fit a needle to its outlet for direct injection.
- Handle samples up to 12mL in volume
- No assembly required
- Replace syringe-coupled devices with single filter units for one-time use
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Available in PES and PVDF
Polyvinylidene Fluoride (PVDF)
Sample typr: Low protein binding. For aqueous samples, samples ≤60% organic solvent
Polyethersulfone (PES)
For aqueous samples, low protein binding
Specifications
0.45 μm | |
140 μL | |
Male slip luer | |
Syringeless Filter |
3 cm2 | |
Polypropylene (PP) | |
No | |
Up to 12 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.