Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Autovial Syringeless Filters::Autovial 12::Without Prefilter Available on GSA/VA Contract for Federal Government customers only.

For removing particulates from potentially hazardous samples

Manufacturer:  GE Healthcare AV125NPUAQU

 View more versions of this product

Catalog No. 09-921-1C


Add to cart

Description

Description

Support stand protects slip-Luer tip, recessed to reduce chance of contamination. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial or fit a needle to its outlet for direct injection.

  • Handle samples up to 12mL in volume
  • No assembly required
  • Replace syringe-coupled devices with single filter units for one-time use
  • Preassembled units consist of a filter barrel and a plunger
  • Built-in air purge minimizes holdup
  • Integrated construction eliminates filter pop-off
  • Available in PES and PVDF

Polyvinylidene Fluoride (PVDF)
Sample typr: Low protein binding. For aqueous samples, samples ≤60% organic solvent
Polyethersulfone (PES)
For aqueous samples, low protein binding

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Syringeless Filter
Sample preparation, Difficult-to-filter samples, Quick filtration of samples, Low nonspecific protein binding membrane for samples in aqueous solutions and/or organic solvents.
140 μl
Up to 12mL
No
Male slip luer
12 mL
PTFE
Polypropylene (PP)
The Whatman Autovial Syringeless Filter replaces multiple syringe-coupled sample filtration devices with a single, disposable unit.
0.45μm
3 cm2
Autovial 12 Syringeless Filter, no prefilter, 0.45 μm, PVDF filtration medium (50 pcs)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit