Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Autovial Syringeless Filters::Autovial 12::Without Prefilter
For removing particulates from potentially hazardous samples
Supplier: Cytiva AV125NPUPSU
Description
Support stand protects slip-Luer tip, recessed to reduce chance of contamination. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial or fit a needle to its outlet for direct injection.
- Handle samples up to 12mL in volume
- No assembly required
- Replace syringe-coupled devices with single filter units for one-time use
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Available in PES and PVDF
Polyvinylidene Fluoride (PVDF)
Sample typr: Low protein binding. For aqueous samples, samples ≤60% organic solvent
Polyethersulfone (PES)
For aqueous samples, low protein binding
Specifications
Autovial 12 Syringeless Filter, no prefilter, 0.45 μm, PES filtration medium (50 pcs) | |
0.45 μm | |
Sample preparation, Difficult-to-filter samples, Quick filtration of samples, Low non-specific protein binding membrane for samples in aqueous solutions. | |
Polypropylene (PP) | |
No | |
Up to 12 mL |
PES | |
3 cm2 | |
140 μL | |
Male slip luer | |
Syringeless Filter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction