Whatman™ Benchkote Plus Surface Protector Sheets Available on GSA/VA Contract for Federal Government customers only.

Prevent many common sources of laboratory contamination, such as those involving hazardous materials on shared workspace benches. Whatman™ Benchkote Plus bench liner can also help with the disposal of hazardous materials.

Manufacturer: Whatman™ 23016150

Catalog No. 12-007-183A

  Check Availability
Add to Cart

    Specifications

    Absorbancy 0.75 l/m2
    Length (Metric) 500mm
    Width (Metric) 600mm
    Quantity 50 Sheets
    Dimensions (L x W) 500 x 600mm
    Product Type Surface protector
    Material Polyethylene
    Length (English) 19 in.
    Width (English) 23 in.
    For Use With (Application) Suitable for saturation with disinfectant

    • Strong: Benchkote Plus bench liner is strong and tear resistant when wet or dry
    • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
    • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
    • Disposable: Benchkote Plus bench paper can be incinerated after use
    • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit