Learn More
Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/B Available on GSA/VA Contract for Federal Government customers only.
Filters fine particles with high wet-strength and high loading capacity. Whatman™ Binder-Free Microfiber Filters, Grade GF/B are particularly useful for liquid clarification and liquid scintillation counting.
Manufacturer:  Whatman™ 1821185
 View more versions of this product
Catalog No. 09-927-765
  Check Availability
Add to cart
Specifications
Description
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500ºC. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.
Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
  • Fine particle filter with high wet-strength and high loading capacity
  • Useful for liquid clarification and liquid scintillation counting
  • Particularly useful when liquid clarification or solids quantification is required for heavily loaded fine particulate suspensions
  • Used in LSC techniques where high loading capacity is required
  • Can be used as a finely retentive membrane filter
Description & Specifications
Specifications
7.2 in.
Circle
1μm
25pack
500°C
675μm
185mm
5.5 sec./100mL
GF/B
Glass microfiber filter
932°F
0.02 in.
143g/m2
Borosilicate Glass