Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Binder-Free Glass Microfiber Filters GF/F Circles

Served as the basis filter for EPA standard for the toxicity characteristic leaching procedure. Whatman™ Binder-Free Glass Microfiber Filters GF/F Circles offers an extremely high loading capacity and flow rate.

$123.00 - $1362.00

Specifications

Autoclavable Autoclavable
Description Whatman Grade GF/F filter from Cytiva's business is manufactured from pure borosilicate glass. GF/F offers very fine particle retention and good loading capacity and flow rate.
Format Circle
Material Borosilicate Glass
Particle Retention 0.7μm
View More Specs

Products 16
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
09-874-60
View Documents
Cytiva
1825021
21 mm
Pack of 100 for $177.00
Edge
Add to cart
 
09-874-62
View Documents
Cytiva
1825024
24 mm
Pack of 100 for $175.00
Add to cart
 
09-874-64
View Documents
Cytiva
1825025
25 mm
Pack of 100 for $169.47
Edge
Add to cart
 
09-874-70
View Documents
Cytiva
1825037
37 mm
Pack of 100 for $227.00
Add to cart
 
09-874-66
View Documents
Cytiva
1825042
42.5 mm
Pack of 100 for $234.00
Edge
Add to cart
 
09-874-71
View Documents
Cytiva
1825047
47 mm
Pack of 100 for $234.68
Edge
Add to cart
 
09-874-68
View Documents
Cytiva
1825055
55 mm
Pack of 100 for $296.00
Add to cart
 
09-874-72
View Documents
Cytiva
1825070
70 mm
Pack of 100 for $343.00
Edge
Add to cart
 
09-874-73
View Documents
Cytiva
1825090
90 mm
Pack of 25 for $123.00
Edge
Add to cart
 
09-874-74
View Documents
Cytiva
1825110
110 mm
Pack of 25 for $190.00
Edge
Add to cart
 
09-874-75
View Documents
Cytiva
1825125
125 mm
Pack of 25 for $214.00
Add to cart
 
09-874-80
View Documents
Cytiva
1825142
142 mm
Pack of 25 for $280.00
Edge
Add to cart
 
09-874-76
View Documents
Cytiva
1825150
150 mm
Pack of 25 for $327.00
Add to cart
 
09-924-544
View Documents
Cytiva
1825257
257 mm
Pack of 25 for $830.00
Add to cart
 
09-874-78
View Documents
Cytiva
1825293
293 mm
Pack of 25 for $1,362.00
Add to cart
 
09-928-057
View Documents
Cytiva
1825015
15 mm
Pack of 100 for $329.00
Add to cart
 
Description

Description

Served as the basis filter for EPA standard for the toxicity characteristic leaching procedure. Whatman™ Binder-Free Glass Microfiber Filters GF/F Circles offers an extremely high loading capacity and flow rate.

Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500ºC. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.

Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
  • High loading capacity
  • 0.6–0.8 µm nominal particle retention
  • Medium filtration speed
Applications:
  • DNA binding and purification
  • Filtering finely precipitated proteins
  • Filtering biochemical solutions, fluids, and nucleic acids
Specifications

Specifications

Autoclavable
Circle
0.7μm
Recommended for DNA binding and purification.Very effective in filtering finely precipitated proteins.Can be used in conjunction with GF/D as a prefilter for the successful clarification of extremely ‘difficult’ biochemical solutions and fluids and nucl
Circle
500°C
420 μm
41 mL/min
Whatman Grade GF/F filter from Cytiva's business is manufactured from pure borosilicate glass. GF/F offers very fine particle retention and good loading capacity and flow rate.
Borosilicate Glass
19 sec./100mL/in.2
Grade GF/F
932°F
0.016 in.
Filter for TCLP Test Use
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit