Learn More
GE Healthcare Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/B Sheets Available on GSA/VA Contract for Federal Government customers only.
Filters fine particles with high wet-strength and high loading capacity. Whatman™ Binder-Free Microfiber Filters, Grade GF/B Sheets are particularly useful for liquid clarification and liquid scintillation counting.
Manufacturer:  GE Healthcare 1821915
 View more versions of this product
Catalog No. 09-874-28A
Add to cart
Description
Description
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500ºC. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.
Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
  • Fine particle filter with high wet-strength and high loading capacity
  • Useful for liquid clarification and liquid scintillation counting
  • Particularly useful when liquid clarification or solids quantification is required for heavily loaded fine particulate suspensions
  • Used in LSC techniques where high loading capacity is required
  • Can be used as a finely retentive membrane filter
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
GE’s Whatman brand Grade GF/B is a thick filter with fine particle filter and good loading capacity. GF/B filters are particularly useful for liquid clarification and liquid scintillation counting.
Sheets
1.0 μm
25 Pack
500°C
675 μm
143g/m2
Borosilicate glass
5.5 sec./100mL
Grade GF/B
Glass microfiber filter
932°F
0.02 in.
81 mL/min
Used in LSC techniques where high loading capacity is required.
Grade GF/B Filter for Liquid Scintillation, 460 x 570 mm sheet (25 pcs)