Learn More
GE Healthcare Whatman™ Cellulose Nitrate Membrane Filters
Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.
Manufacturer:  GE Healthcare 10401306
 View more versions of this product
Catalog No. 09-927-367
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Among the strongest of their type available, as measured and compared by burst pressure tests.
  • Narrow pore size distribution for improved surface capture and analysis.
  • Low levels of extractables to ensure sample integrity.
  • High strength and flexibility
  • Increased temperature stability
  • Reduced shrinkage
Specifications
Specifications
100Pack
1.9 in.
105 to 140μm
Circle
25mm
Cellulose Nitrate
0.2μm