Learn More
GE Healthcare Whatman™ Cellulose Nitrate Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.
Manufacturer:  GE Healthcare 10401312
 View more versions of this product
Catalog No. 09-927-370
Add to cart
Description
Description
Among the strongest of their type available, as measured and compared by burst pressure tests.
  • Narrow pore size distribution for improved surface capture and analysis.
  • Low levels of extractables to ensure sample integrity.
  • High strength and flexibility
  • Increased temperature stability
  • Reduced shrinkage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100Pack
1.85 in.
105 to 140μm
Circle
47mm
Cellulose Nitrate
0.2μm