Learn More
GE Healthcare Whatman™ Drying Rack for 903 Protein Saver Cards
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
Manufacturer:  GE Healthcare 10537173
 View more versions of this product
Catalog No. 09-800-963
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • For use with neonatal human diagnostic testing cards
  • CE Marked
  • Accommodate multiple collection forms at one time
Specifications
Specifications
903 Protein Saver Card
Mounting Sticker not included
10 Pack
Rack, Drying