Learn More
GE Healthcare Whatman™ Elutrap™ Electroelution Membranes Available on GSA/VA Contract for Federal Government customers only.
Designed to isolate nucleic acids and proteins from agarose or polyacrylamide gel slices
Manufacturer:  GE Healthcare 10404090
 View more versions of this product
Catalog No. 09-301-141
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • For most horizontal gel electrophoresis chambers
  • Efficient current flow
  • Handles up to four samples simultaneously
  • Purifies nucleic acids 14bp to 150Kb; proteins larger than 3 to 5kD
  • No salt cushions or special buffers required
  • Adjustable trap allows optimization of final sample volume
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Elutrap™ electroelution system
BT1 membrane
Eluting nucleic acids (14 bp to 150Kb) and proteins (larger than 3 to 5kD) from gel slices