Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Filter Funnel Plate, Polypropylene
Use with Whatman™ 3-piece Filter Funnels. Whatman™ Filter Funnel Plate, Polypropylene is an optional extra plate recommended for most acids.
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 1950109
Catalog No. 09-877-8
Add to cart
Description
Description
  • Polypropylene
  • Recommended for most acids (except concentrated nitric acid and fuming sulfuric acid) at room temperature
  • Also suitable for most alcohols, glycols, ethers and ketones
  • Maximum working temperature: 100°C
Specifications
Specifications
3.5 in.
212°F
Recommended for most acids and also suitable for most alcohols, glycols, ethers and ketones
Polypropylene
9cm
100°C
Whatman™ 3-piece Filter Funnel