Learn More
Cytiva Whatman™ Filter Funnel Replacement Reservoir
$190.10 - $194.40
Specifications
Description Replacement filter funnel reservoir
For Use With (Equipment) 1950 Series 3-Piece Filter Funnels for glass microfiber filters
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Inside Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Inside Price Quantity & Availability  
09-927-621
View Documents
Cytiva
1950217
210 mL 70 mm
Pack for $190.10
Add to cart
09-927-620
View Documents
Cytiva
1950209
200 mL 90 mm
Pack for $194.40
Add to cart
Description
Description
Specifications
Specifications
Replacement filter funnel reservoir
1950 Series 3-Piece Filter Funnels for glass microfiber filters