Whatman™ FilterCup Vacuum Filter Funnels
Manufacturer: Whatman™ 1600-003
 View all options for this product
Catalog No. 09-878C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Quantity (No. per Case) 25 Pack
    Material Polypropylene
    Product Type Vacuum Filter Funnel
    Disposable Yes
    Filtration Area 31cm2
    For Use With (Application) Aqueous solutions, dilute acids, alkalis and a range of organic solvents
    Material (Membrane) Cellulose
    Volume (Metric) 250mL
    • FilterCup device has 250mL capacity and a 70mm diameter membrane available in cellulose or glass microfiber in several filter grades (see chart)
    • Recommended for aqueous solutions, dilute acids, alkalis and a range of organic solvents
    • Usually used under vacuum with the FilterCup stem (Cat. No. 09-878N, sold separately)
    • FilterCup units easily snap into and out of the stem
  • Description & Specifications
Related Products
Product Content Feedback