Whatman™ FilterCup Vacuum Filter Funnels
Manufacturer: Whatman™ 1600820
 View more versions of this product
Catalog No. 09-878H
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Vacuum Filter Funnel
    Material Polypropylene
    Disposable Yes
    Filtration Area 31cm2
    For Use With (Application) Aqueous solutions, dilute acids, alkalis and a range of organic solvents
    Material (Membrane) Glass Microfiber
    Volume (Metric) 250mL
    Quantity (No. per Case) 25 Pack
  • Description & Specifications
Related Products
Product Content Feedback