Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Grade 589/2 Quantitative Filter Paper Available on GSA/VA Contract for Federal Government customers only.

Retains medium fine precipitates using material with virtually no ash content. Whatman™ Grade 589/2 Quantitative Filter Paper is used when medium fine precipitates are being collected and papers must be ashed to support analysis.

Manufacturer:  GE Healthcare 10300114

 View more versions of this product

Catalog No. 09-924-115


Add to cart

Description

Description

Designed for gravimetric analysis

  • White Ribbon Filter for medium fine precipitates
  • Ashless
  • Medium filtration speed
  • For sand content measurement in foodstuffs
  • For flour grade determination
  • For aqueous suspension analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Grade 589/2 Medium Fine Precipitate Filter, 185 mm circle (100 pcs)
Whatman Grade 589/2 filters from GE are used in routine quantitative analyses, including analysis of aqueous suspension in paper, flour analysis, and sand content quantification.
Grade 589/2
Folded
185 mm
Medium
Ashless
100Pack
High-quality cotton linters
180μm
4 to 12μm
White
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit