Whatman™ FTA™ DMPK Cards

DBS microvolume sampling requires only 10 to 20µL per sample

Manufacturer: Whatman™ WB120222

 View all options for this product

Catalog No. 09-927-967


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    For Use With (Application) For protein-based biomolecules, on-substrate cleanup has potential to convey greater analyte stability, requires fewer experimental animals
    Format Card
    Formulation Type A, Indicating
    Product Type FTA DMPK Card

    • Fewer experimental animals are required — Toxicokinetic data are derived from Toxicology animals, thereby eliminating satellite colonies
    • More consistent data are obtained through more serial sampling from individual animals and less reliance on composite data
    • 3-Step DBS procedure is much more straightforward than the cumbersome centrifugation, isolation and cleanup of plasma
    • On-substrate cleanup has the potential to convey greater analyte stability, especially for enzyme-sensitive compounds.
    • Room temperature stability saves on cost of dry ice shipments
    • Shipping without dry ice allows more remote sampling

    • Card types A, B and C available, as well as starter packs
    • Both non-indicating and indicating cards available
    • FTA DMPK-C card is not impregnated with chemicals and may be better suited for protein-based biomolecules

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit