Whatman™ Indicating FTA Classic Cards

Lyses cells on contact, denatures proteins and protects DNA from degradation. Whatman™ Indicating FTA Classic Cards includes a pink dye that turns white when a clear sample is applied.

Manufacturer: Whatman™ WB120315

 View more versions of this product

Catalog No. 09-302-195

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Includes Pink indicating dye
    For Use With (Application) Clear samples, such as buccal cells or saliva
    Product Type Indicating
    Volume (Metric) Sample 500μL max. per card
    No. of Samples 4
    Format Four sample areas
    Sample Size 125μL per sample area
    For Use With (Equipment) Direct STR amplification
    Quantity 250 Pack,250 Pack
    Product Name Indicating FTA Classic card

    Collect, stabilize, process, transport, and archive buccal DNA samples.

    • Improves process efficiency by enabling direct amplification for STR analysis due to immediate DNA capture on the FTA card.
    • Includes an inert dye, which turns to white when a buccal sample or saliva is added for easy confirmation of sample addition.
    • Immediate stabilization and protection of nucleic acids for long term archiving of samples at room temperature.
    • Different card options to support manual or automated processing for versatility and incorporation into current workflows.
    • DNA from blood stored at room temperature on FTA cards exhibits no loss in PCR efficiency after 22 years and counting
    • Include a pink indicating dye that turns to white when a clear sample is applied.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit