Learn More
GE Healthcare Whatman™ FTA™ Nucleic Acid Collection, Storage and Purification Cards: FTA Gene
Collect, store, and transport DNA at room temperature
Manufacturer:  GE Healthcare WB120314
 View more versions of this product
Catalog No. 09-302-194
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Patented FTA technology simplifies the isolation, storage, and processing of nucleic acids from biological sources including whole blood, buccal cells, and plant material. Sample is applied to the FTA card and allowed to dry; the cells are lysed on contact. The released DNA is captured, stabilized, and protected from nucleases, oxidation, and UV damage.
  • DNA from blood stored at room temperature on FTA cards exhibits no loss in PCR efficiency after 22 years and counting
  • FTA Gene Card is housed in a chipboard frame for compatibility with most automatic dispensing/pipetting systems when used with the FTA Gene Card Tray (09-923-349)
Specifications
Specifications
Forensics, transgenic identification, transfusion medicine/HLA typing, plasmid screening, food and agriculture testing, drug discovery, genomics, STR analysis, animal identification, whole genome amplification
Non-Indicating FTA Gene Card
75μL