Learn More
GE Healthcare Whatman™ FTA™ Nucleic Acid Collection, Storage and Purification Cards: FTA Mini
Collect, store, and transport DNA at room temperature
Manufacturer:  GE Healthcare WB120317
 View more versions of this product
Catalog No. 09-302-197
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Patented FTA technology simplifies the isolation, storage, and processing of nucleic acids from biological sources including whole blood, buccal cells, and plant material. Sample is applied to the FTA card and allowed to dry; the cells are lysed on contact. The released DNA is captured, stabilized, and protected from nucleases, oxidation, and UV damage.
  • DNA from blood stored at room temperature on FTA cards exhibits no loss in PCR efficiency after 22 years and counting
  • FTA cards are available in white and as pink indicating cards for colorless fluids
  • Pink indicating cards disclose where colorless samples have been spotted by changing to white spot
Specifications
Specifications
Forensics, transgenic identification, transfusion medicine/HLA typing, plasmid screening, food and agriculture testing, drug discovery, genomics, STR analysis, animal identification, whole genome amplification
Indicating FTA Mini card
125μL