Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ GD/X Nylon Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X Nylon Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
$621.00 - $4881.00
Specifications
Housing Material Pigment free polypropylene (PP)
Inlet Style Female luer lock
Outlet Style Male luer
Volume (Metric) ≤10mL
Disposable Yes
View More Specs
Products 9
Catalog Number Mfr. No. Diameter (Metric) Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Pore Size No. per Pack Price Quantity & Availability  
09-927-26A
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68701302
13mm 0.2μm 150/pk.
Pack of 150 for $621.00
Add to cart
09-927-26B Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68701304
13mm 0.45μm 150/pk.
Pack of 150 for $621.00
Add to cart
09-927-27B
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68711304
13mm 0.45μm 1500/pk.
Pack of 1500 for $3,301.00
Add to cart
09-927-27A
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68711302
13mm 0.2μm 1500/pk.
Pack of 1500 for $3,373.00
Add to cart
09-927-27C
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68712502
25mm 0.2μm 1500/pk.
Pack of 1500 for $4,881.00
Add to cart
09-927-26C Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68702502
25mm 0.2μm 150/pk.
Pack of 150 for $805.00
Add to cart
09-927-26D Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68702504
25mm 0.45μm 150/pk.
Pack of 150 for $805.00
Add to cart
09-927-27D
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68712504
25mm 0.45μm 1500/pk.
Pack of 1500 for $4,881.00
Add to cart
09-927-48
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
68702550
25mm 5.0μm 150/pk.
Pack of 150 for $805.00
Add to cart
Description
Description
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X Nylon Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
Constructed of a polypropylene housing with a prefiltration stack of Whatman GMF 150 (graded density) and GF/F glass microfiber prefilters. This design prevents the buildup of back pressure typically caused by the blocking of an unprotected membrane.
  • Syringe filter with integrated prefiltration
  • Female Luer-Lok™ inlet; male slip Luer outlet
  • Flow should enter from the inlet
  • Autoclavable at 121°C (250°F) at 15psi for 20 min.
  • Maximum temperature, 131°C (267°F) Filtration area: 13mm - 1.3cm2; 25mm - 4.6cm2 Pressure limit: 13mm - 100psi; 25mm - 75psi
  • All materials biosafe—pass USP Class VI requirements
  • Prefiltration media: 100% Borosilicate glass fiber GMF 150 10µm: 1µm GF/F 0.7µm
  • Disposable filtration devices provide labor saving efficiency while ensuring consistent filter performance
  • High loading capacity for samples with high solids content
  • Suitable for capillary electrophoresis, HPLC, and UV/VIS sample preparation
Nonsterile GD/X filters are available in two diameters: 13 mm and 25 mm, and three pore sizes: 0.2 µm, 0.45 µm, and 5 µm
Order Info
Each unit includes a stack of Whatman Multigrade GMF 150 (10:1µm) and Grade GF/F 0.7µm prefilters
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Pigment free polypropylene (PP)
Male luer
Yes
Female luer lock
≤10mL