Learn More
Whatman™ GD/XP Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Incorporates a two-layer polypropylene prefilter to minimize levels of ion extractables. Whatman™ GD/XP Syringe Filters are suitable  for samples that require inorganic ion analysis.
Manufacturer:  Whatman™ 69712504
 View more versions of this product
Catalog No. 09-930-21
  Check Availability
Add to cart
Specifications
Description
Designed to prepare and enhance inorganic ion analysis of hard-to-filter samples. Recommended for analyzing ion content that would otherwise experience interference from glass-based extractables
  • 25mm diameter with 0.45µm pore size
  • Proprietary polypropylene prefiltration stack
  • Autoclave at 121°C at 1 bar (15psi) for 20 minutes
  • Nonsterile
  • Extremely low protein binding filter
  • High loading volume for filtration of challenging samples
  • Use for trace metal analysis; particularly ion analysis using inductively coupled plasma-mass spectrometry (ICP-MS)
Description & Specifications
Specifications
Syringe Filter
Nylon (NYL)
Polypropylene
>10mL
1500/Pk.
25mm
Trace metal analysis
250uL (1.4mL)
0.45μm