Learn More
Cytiva Whatman™ Glass Fiber Filter Papers - Grade 31
Offers fine particle retention and high flow rate, as well as good loading capacity
Manufacturer:  Cytiva 10372803
Catalog No. 09-927-276
Add to cart
Description
Description
Specifications
Specifications
Glass Fiber
Fine particle retention and high flow rate, as well as good loading capacity
2.1 in.
Fast
31
5.5 cm