Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Whatman™ Filter Funnel Plates, PTFE Available on GSA/VA Contract for Federal Government customers only.

Use with Whatman™ 3-piece Filter Funnels. Whatman™ Filter Funnel Plate, PTFE is an optional extra plate ideal for acids, alkalis and solvents.

Manufacturer:  Whatman™ 1950117

 View more versions of this product

Catalog No. 09-927-617


  Check Availability
Add to cart

Specifications

Description

 

  • Maximum working temperature, 200°C (392°F)
  • Ideal for virtually all common acids, alkalis and solvents at temperatures up to 100°C
  • Maximum working temperature: 200°C
  •  

Description & Specifications

Specifications

70mm
<200°C
PTFE
<392°F
Whatman 1950 series filter funnels
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit