Learn More
GE Healthcare Whatman™ Glass Microfiber Filter Paper Grade 9 Available on GSA/VA Contract for Federal Government customers only.
Glass microfiber filter paper
Manufacturer:  GE Healthcare 10370202
 View more versions of this product
Catalog No. 09-924-274
Add to cart
Description
Description
  • Filters used for various applications from fine and coarse particle analysis to prefilter membranes for crop protection residues
  • Flow rate: 27 s/100mL/1.56cm2
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
50mm
GF 9
27 s/100mL/1.56cm2
350μm
2 in.
Glass Microfiber
Circle