Learn More
Cytiva Whatman™ Glass Microfiber Filters with Organic Binder, Grade GF 10
For use as a weighing aid for infrared weighing, and as a roll filter in automatic air filtration units
Supplier:  Cytiva 10370391
Catalog No. 09-920-093
Add to Cart
Description
Description
Specifications
Specifications
Roll
Weighing Aid for Infrared Weighing
1 Each
Filter
42 m
Glass Membrane, Organic Binder
Grade GF 10
180°C
42 m x 60 mm
60 mm
Product Suggestions
Product Suggestions