Learn More
GE Healthcare Whatman™ Grade 0858 Filter Papers for Technical Use Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  GE Healthcare 10334365
 View more versions of this product
Catalog No. 09-924-354
Add to cart
Description
Description
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0858
4 in.
58cm
Medium
170μm
11cm
Super-refined Cellulose
23 in.
4 x 23 in. (11 x 58cm)