Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ Grade 114 Wet-Strengthened Qualitative Filter Paper: Circle

Coarse paper with creped or smooth surface

Manufacturer:  GE Healthcare 1114150

 View more versions of this product

Catalog No. 09-833G


Add to cart

Description

Description

  • Have high wet strength due to the addition of a small quantity of chemically stable resin

Cat. No. 09-833F is made to order, and therefore not returnable

Specifications

Specifications

100 Pieces
Only half the thickness of Grade 113 and suitable for coarse or gelatinous precipitates. Smooth surface for easy recovery of precipitates.
Grade 114
Use for coarse particles and gelatinous precipitates.
Circle
Grade 114 Qualitative Filter Paper Wet-Strengthened, circle, 150 mm
150 mm
Cellulose
25μm
190 μm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit