Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Grade 2294 Qualitative Filter Paper

Made from super-refined cellulose and specifically designed for filtration application. Whatman™ Grade 2294 Qualitative Filter Paper has uses that range from beverage filtration to the purification of electroplating baths.

$65.30 - $210.50

Specifications

Format Circle
Grade Grade 2294
Material Cellulose
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
09-924-370
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10342810
110 mm
Pack of 100 for $210.50
Add to cart
 
09-924-371
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10342860
180 mm
Pack of 100 for $65.30
Add to cart
 
09-924-372
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10342862
210 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Very thick filter card with high wet strength
  • Very high flow rate
  • Retains medium to coarse particles

Specifications

Specifications

Circle
Grade 2294
Cellulose
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit