Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Grade 287 1/2 Folded Filter Paper
Folded filter paper saves time, increases loading capacity
$83.00 - $100.00
Specifications
Filtration Speed | Approx. 330 s |
---|---|
For Use With (Application) | For clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry |
Grade | Grade 287 ½ |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
09-924-143
|
Cytiva
10310247 |
185 mm |
Pack of 50 for $100.00
|
|
|||||
09-924-142
|
Cytiva
10310245 |
150 mm |
Pack of 50 for $83.00
|
|
|||||
Description
- Kieselguhr paper with medium to slow flow
- For separation of very fine semi-colloidal turbidity
- For clarifying milk serum, starch or sugar-coated solutions, soil suspensions
Specifications
Approx. 330 s | |
For clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry | |
Grade 287 ½ |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title