Learn More
GE Healthcare Whatman™ Grade 287 1/2 Folded Filter Paper Available on GSA/VA Contract for Federal Government customers only.
Folded filter paper saves time, increases loading capacity
Manufacturer:  GE Healthcare 10310245
 View more versions of this product
Catalog No. 09-924-142
Add to cart
Description
Description
  • Kieselguhr paper with medium to slow flow
  • For separation of very fine semi-colloidal turbidity
  • For clarifying milk serum, starch or sugar-coated solutions, soil suspensions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Grade 287 ½ Qualitative Filter Paper Folded (Prepleated), 150 mm
150 mm
Grade 287 ½
Medium Slow
Kieselguhr prepleated paper with a medium to slow flow rate and additional adsorption effect.
150mm
For clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry
Folded