Learn More
GE Healthcare Whatman™ Grade 520a Qualitative Folded Paper Filter Circle
For the filtration of viscous liquids and emulsions such as sweetened juices, spirits,syrups, resin solutions, oils or plant extracts
Manufacturer:  GE Healthcare 10331451
 View more versions of this product
Catalog No. 09-924-343
Add to cart
Description
Description
  • Great wet strength
  • Very high flow rate
  • Super-refined cellulose
Specifications
Specifications
Grade 520 a ½ Qualitative Filter Paper Folded (Prepleated), 240 mm
240 mm
Cellulose
18 μm
Circle
A thin paper with great wet strength and a very high flow rate. Frequently used in technical applications such as the filtration of viscous liquids and emulsions (e.g., sweetened juices, spirits and syrups, resin solutions, oils, or plant extracts).
Grade 520 a ½
Frequently used in technical applications such as the filtration of viscous liquids and emulsions.
High