Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Circles

High wet strength due to the addition of chemically stable resin

Manufacturer: Whatman™ 1091190

 View more versions of this product

Catalog No. 09-927-508

  Check Availability
Add to Cart

    Specifications

    Quantity 1000 Pieces (subdivided into 100)
    For Use With (Application) Used to assay sucrose in cane sugar
    Pore Size (Metric) 10μm
    Particle Retention 10μm
    Format Circle
    Thickness 205μm
    Grade 91
    Material Cellulose
    Diameter (Metric) 190mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit