Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Circles
High wet strength due to the addition of chemically stable resin
Supplier: Cytiva 1091150
Description
High wet strength due to the addition of chemically stable resin
- Wet strengthened grade
- General purpose creped filter
- For less critical routine analysis
- Used to assay sucrose in cane sugar
Specifications
Grade 91 Qualitative Filter Paper Wet-Strengthened, circle, 150 mm (subdivided into 100) | |
Cellulose | |
10 μm | |
A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration. | |
1000 Pieces (subdivided into 100) | |
Qualitative Filter Paper | |
5.9 in. |
Disc | |
10 μm | |
70 sec. | |
Grade 91 | |
205 μm | |
Ø 150 mm | |
150 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction