Learn More
Whatman™ Grade 93 Wet Strengthened Qualitative Filter Papers - Sheets
High wet strength due to the addition of chemically stable resin
Manufacturer:  Whatman™ 1093935
 View more versions of this product
Catalog No. 09-927-531
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
500 Pieces
93
24 in.
610mm
10μm
Sheet
145μm
125 mm
610mm
Cellulose
24 in.
10μm
24 x 24 in. (610 x 610mm)