Learn More
Cytiva Whatman Grade 934-AH RTU Glass Microfiber Filters
Ready to use, Grade 934-AH RTU Glass Microfiber Filter Papers are the time-saving version of GE’s 934-AH glass microfiber filter papers, which are widely used for total suspended solids analysis.
$126.00 - $334.00
Specifications
Autoclavable Not Autoclavable
Material Glass Microfiber
Particle Retention 1.5 μm
Sterility Non-sterile
Grade 934-AH RTU
View More Specs
Products 14
Catalog Number Mfr. No. Diameter (Metric) Type Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Type Price Quantity & Availability  
09-928-043
View Documents
Cytiva
4827047
47 mm Filter for volatiles, pre-rinsed and ignited
Pack of 100 for $164.00
Edge
Add to Cart
09-928-044
View Documents
Cytiva
4827070
70 mm Filter for volatiles, pre-rinsed and ignited
Pack of 100 for $212.00
Add to Cart
09-928-045
missing translation for 'viewDocuments'
Cytiva
4827090
90 mm Filter for volatiles, pre-rinsed and ignited
Pack of 100 for $231.00
Add to Cart
09-928-036
missing translation for 'viewDocuments'
Cytiva
3827035
35 mm RTU Filter for volatile, pre-weighed
Pack of 100 for $156.00
Add to Cart
09-928-049
missing translation for 'viewDocuments'
Cytiva
9907070
70 mm RTU Filter for solids
Pack of 100 for $212.00
Add to Cart
09-928-037
missing translation for 'viewDocuments'
Cytiva
3827042
42.5 mm RTU Filter for volatile, pre-weighed
Pack of 100 for $156.00
Add to Cart
09-928-038
missing translation for 'viewDocuments'
Cytiva
3827047
47 mm RTU Filter for volatile, pre-weighed
Pack of 100 for $173.00
Add to Cart
09-928-046
missing translation for 'viewDocuments'
Cytiva
9927047
47 mm RTU Filter for solids, double weigh
Pack of 100 for $258.00
Add to Cart
09-928-039
missing translation for 'viewDocuments'
Cytiva
3827070
70 mm RTU Filter for volatile, pre-weighed
Pack of 100 for $225.00
Add to Cart
09-928-047
missing translation for 'viewDocuments'
Cytiva
9927070
70 mm RTU Filter for solids, double weigh
Pack of 100 for $310.00
Add to Cart
09-928-040
missing translation for 'viewDocuments'
Cytiva
3827090
90 mm RTU Filter for volatile, pre-weighed
Pack of 100 for $260.00
Add to Cart
09-928-048
missing translation for 'viewDocuments'
Cytiva
9927090
90 mm RTU Filter for solids, double weigh
Pack of 100 for $334.00
Add to Cart
09-928-041
missing translation for 'viewDocuments'
Cytiva
4827035
35 mm Filter for volatiles, pre-rinsed and ignited
Pack of 100 for $126.00
Add to Cart
09-928-042
missing translation for 'viewDocuments'
Cytiva
4827042
42.5 mm Filter for volatiles, pre-rinsed and ignited
Pack of 100 for $133.00
Add to Cart
Description
Description
  • Grade 934-AH RTU Glass microfiber filter papers have been pre-washed, dried (or ignited), cooled, desiccated and weighed, eliminating pre-treating steps required by US EPA Laboratory Standard Method 2540 parts C, D, and E
  • Fine particle retention of 1.5μm
  • High flow rates to process heavy, turbid samples
  • Packaged with a barcoded aluminum pan noting the filter weight which can be used to find documentation of the filter weight
  • 934-AH RTU family comes in three different varieties: 934-AH RTU for Suspended and Dissolved Solids; RTU for Volatiles; RTU Double Weigh
  • Economy versions of 934-AH RTU dissolved, suspended, and volatiles testing filters are also available
  • Economy versions are are pre-washed and dried but not weighed and do not come with barcodes
Applications include dissolved, suspended and volatiles solid analysis; also wastewater monitoring applications including rivers, lakes and coastal waters, as well as analysing purified discharge of wastewater treatment plants and monitoring discharge water from industrial plants
Specifications
Specifications
Not Autoclavable
1.5 μm
934-AH RTU
341 mL/min
Glass Microfiber
Non-sterile
100
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.