Learn More
Cytiva Whatman™ Grade 934-AH RTU Glass Microfiber Filters, Ready to Use PROMO
Widely used for total suspended solids analysis.
Manufacturer:  Cytiva 29822127
Catalog No. 14-380-935LR2
Add to cart
Description
Description
  • Prewashed, preweighed RTU filters eliminate pretreating steps otherwise required by Standard Method 2540 parts C, D, and E.
  • Fine particle retention of 1.5 μm at high flow rates
  • Able to process heavy, turbid samples
Ready to use wastewater filter Whatman 934-AH RTU filters meet the requirements for total suspended solids analysis of water according to the latest edition of Standard Method 2540D. 934-AH RTU is used as a wastewater filter for a wide range of water monitoring applications including:
  • Monitoring rivers, lakes, and coastal waters
  • Analyzing purified discharge of wastewater treatment plants
  • Monitoring discharge water from industrial plants
RTU filters are packaged within an aluminum pan with the filter weight clearly printed on a heat-resistant label. Each pan has a unique barcode.
Specifications
Specifications
3.7 sec./100 mL/in.2
Glass Microfiber
1.5 μm
550°C
341 mL/min.
64 g/m2
Borosilicate Glass Microfiber
934-AH RTU
Filter
47 mm