Learn More
Cytiva Whatman™ Grade 934-AH RTU Glass Microfiber Filters, Ready to Use PROMO
Ready to use, Grade 934-AH RTU Glass Microfiber Filter Papers are the time-saving version of GE’s 934-AH glass microfiber filter papers, which are widely used for total suspended solids analysis.
$64.89 - $172.80
Specifications
Autoclavable Not Autoclavable
Material Glass Microfiber
Particle Retention 1.5 μm
Sterility Non-sterile
Grade 934-AH RTU
View More Specs
Products 13
Catalog Number Mfr. No. Type Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Type Diameter (Metric) Price Quantity & Availability  
09-928-036LR2
View Documents Promotion Details
Cytiva
29822128
RTU Filter for volatile, pre-weighed 35 mm
Pack of 100 for $79.77
Add to cart
09-928-037LR2
View Documents Promotion Details
Cytiva
29822129
RTU Filter for volatile, pre-weighed 42.5 mm
Pack of 100 for $79.77
Add to cart
09-928-038LR2
View Documents Promotion Details
Cytiva
29822130
RTU Filter for volatile, pre-weighed 47 mm
Pack of 100 for $89.78
Add to cart
09-928-039LR2
View Documents Promotion Details
Cytiva
29822131
RTU Filter for volatile, pre-weighed 70 mm
Pack of 100 for $114.93
Add to cart
09-928-040LR2
View Documents Promotion Details
Cytiva
29822122
RTU Filter for volatile, pre-weighed 90 mm
Pack of 100 for $133.86
Add to cart
09-928-041LR2
View Documents Promotion Details
Cytiva
29822133
Filter for volatile, pre-rinsed and ignited 35 mm
Pack of 100 for $64.89
Add to cart
09-928-042LR2
View Documents Promotion Details
Cytiva
29822134
Filter for volatile, pre-rinsed and ignited 42.5 mm
Pack of 100 for $67.60
Add to cart
09-928-043LR2
View Documents Promotion Details
Cytiva
29822135
Filter for volatile, pre-rinsed and ignited 47 mm
Pack of 100 for $82.94
Add to cart
09-928-044LR2
View Documents Promotion Details
Cytiva
29822136
Filter for volatile, pre-rinsed and ignited 70 mm
Pack of 100 for $108.16
Add to cart
09-928-045LR2
View Documents Promotion Details
Cytiva
29822137
Filter for volatile, pre-rinsed and ignited 90 mm
Pack of 100 for $118.98
Add to cart
09-928-046LR2
View Documents Promotion Details
Cytiva
29822138
RTU Filter for solids, double weigh 47 mm
Pack of 100 for $132.51
Add to cart
09-928-047LR2
View Documents Promotion Details
Cytiva
29822139
RTU Filter for solids, double weigh 70 mm
Pack of 100 for $159.99
Add to cart
09-928-048LR2
View Documents Promotion Details
Cytiva
29822123
RTU Filter for solids, double weigh 90 mm
Pack of 100 for $172.80
Add to cart
Description
Description
  • Grade 934-AH RTU Glass microfiber filter papers have been pre-washed, dried (or ignited), cooled, desiccated and weighed, eliminating pre-treating steps required by US EPA Laboratory Standard Method 2540 parts C, D, and E
  • Fine particle retention of 1.5 μm
  • High flow rates to process heavy, turbid samples
  • Packaged with a barcoded aluminum pan noting the filter weight which can be used to find documentation of the filter weight
  • 934-AH RTU family comes in three different varieties: 934-AH RTU for Suspended and Dissolved Solids; RTU for Volatiles; RTU Double Weigh
  • Economy versions of 934-AH RTU dissolved, suspended, and volatiles testing filters are also available
  • Economy versions are are pre-washed and dried but not weighed and do not come with barcodes
Applications include dissolved, suspended and volatiles solid analysis; also wastewater monitoring applications including rivers, lakes and coastal waters, as well as analysing purified discharge of wastewater treatment plants and monitoring discharge water from industrial plants
Specifications
Specifications
Not Autoclavable
1.5 μm
934-AH RTU
341 mL/min.
Glass Microfiber
Non-sterile
100 Pk.