Learn More
GE Healthcare Whatman™ Grade 995 General Purpose Filter Paper - Circle
Educational grade filter paper recommended for general filtrations
Manufacturer:  GE Healthcare 10318614
 View more versions of this product
Catalog No. 09-924-331
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Smooth-surfaced wood pulp
  • High wet-strength
  • Medium flow strength
Specifications
Specifications
185mm
Cellulose
8 to 12μm
Circle
995
General purpose and non-critical laboratory filtration procedures
Medium