Learn More
GE Healthcare Whatman™ Grade GB004 Blotting Paper
Thick gel blotting paper used for wicking purposes only
Manufacturer:  GE Healthcare 10427904
 View more versions of this product
Catalog No. 09-301-407
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Pure cellulose—produced from the highest quality cotton linters with no additives to ensure that no contamination will occur during transfers
  • Manufactured and tested specifically for blotting applications
  • Provides higher absorbency and more consistent wicking than paper towels
  • Thickness: 1.0mm
  • Ideal for applications where fewer layers of blotting paper, which reduce the risk of trapping air bubbles, must still ensure a high capacity
  • Recommended for capillary blotting of nucleic acids
  • Precisely cut to the most common gel and transfer membrane sizes, eliminating sheet-to-sheet variation