Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman Grade GF/C Glass Microfiber Filters, Binder Free, Ready to Use
Grade GF/C glass filters offer fine particle retention, good flow rate and loading capacity, and support total suspended solids analysis.
Supplier: Cytiva 3822090
Description
- Good flow rate and loading capacity
- Fine nominal particle retention of 1.2μm
- Binder free
- Applications include total suspended solids analysis, industrial waste analysis and potable water
- GF/C RTU pre-treats traditional GF/C filter with washing, drying, cooling (in a desiccator), and weighing steps aligned to EN 872
- Certified weight loss of less than 0.017mg/cm2 after preparation
- Each RTU filter is placed in an individually barcoded aluminum pan that notes the filter weight
Specifications
6.7s/100mL/in2 | |
Borosilicate Glass | |
1.2 μm | |
Grade GF/C | |
550°C | |
RTU Filter, pre-weighed | |
9.0 cm |
GE’s ready-to-use Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis with a fine nominal particle retention of 1.2 μm. | |
1.2 μm | |
Widely used for cell harvesting, liquid scintillation counting and binding assays where more loading capacity is required.Highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is | |
100 Pieces | |
260 μm | |
105 mL/min |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction